COM448612581 Combi-Blocks, Inc.

2,2-Dichloro-N-(4-fluoro-2-nitrophenyl)acetamide

  • MDL number MFCD27981295
  • Cas Number : 1820608-52-3
  • Molecular Weight : 267.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3530
Availability details for
Quantity
  • Availability
  • Add to Cart