COM448612584 Combi-Blocks, Inc.

3-Nitro-N-(2,2,2-trifluoroethyl)benzamide

  • MDL number MFCD06675043
  • Cas Number : 26930-22-3
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3537
Availability details for
Quantity
  • Availability
  • Add to Cart