COM448612589 Combi-Blocks, Inc.

3-(2-Nitrophenylamino)-3-phenylpropan-1-ol

  • MDL number MFCD27981297
  • Cas Number : 1820603-74-4
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3546
Availability details for
Quantity
  • Availability
  • Add to Cart