COM448612591 Combi-Blocks, Inc.

2-Methoxy-4-nitrobenzamide

  • MDL number MFCD12913504
  • Cas Number : 62726-03-8
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3550

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3550
Availability details for
Quantity
  • Availability
  • Add to Cart