COM448612595 Combi-Blocks, Inc.

2-Acetamido-6-methoxybenzoic acid

  • MDL number MFCD24481071
  • Cas Number : 50868-77-4
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3555
Availability details for
Quantity
  • Availability
  • Add to Cart