COM448612596 Combi-Blocks, Inc.

1-Chloro-3-fluoro-2-methyl-4-nitrobenzene

  • MDL number MFCD28014023
  • Cas Number : 1805522-74-0
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3556

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3556
Availability details for
Quantity
  • Availability
  • Add to Cart