COM448612601 Combi-Blocks, Inc.

5-Isopropoxy-2-nitrophenol

  • MDL number MFCD28024834
  • Cas Number : 855839-37-1
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3572

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3572
Availability details for
Quantity
  • Availability
  • Add to Cart