COM448612604 Combi-Blocks, Inc.

5-Chloro-4-methyl-2-nitrophenol

  • MDL number MFCD19374209
  • Cas Number : 100278-74-8
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3575

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3575
Availability details for
Quantity
  • Availability
  • Add to Cart