COM448612606 Combi-Blocks, Inc.

4-(Benzyloxy)-2-nitrophenol

  • MDL number MFCD03926344
  • Cas Number : 96315-18-3
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3577
Availability details for
Quantity
  • Availability
  • Add to Cart