COM448612608 Combi-Blocks, Inc.

2-Amino-5-isopropoxyphenol

  • MDL number MFCD16997362
  • Cas Number : 1243280-86-5
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3579

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3579
Availability details for
Quantity
  • Availability
  • Add to Cart