COM448612612 Combi-Blocks, Inc.

2-Amino-5-chloro-4-methylphenol

  • MDL number MFCD19374213
  • Cas Number : 1267409-17-5
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3583
Availability details for
Quantity
  • Availability
  • Add to Cart