COM448612614 Combi-Blocks, Inc.

2-Amino-4-(benzyloxy)phenol

  • MDL number MFCD24646652
  • Cas Number : 102580-07-4
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3586
Availability details for
Quantity
  • Availability
  • Add to Cart