COM448612615 Combi-Blocks, Inc.

2-Amino-5-chloro-4-fluorophenol

  • MDL number MFCD19374214
  • Cas Number : 1191063-34-9
  • Molecular Weight : 161.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3587

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3587
Availability details for
Quantity
  • Availability
  • Add to Cart