COM448612619 Combi-Blocks, Inc.

(S)-2-Amino-1-phenylethanol hydrochloride

  • MDL number MFCD11114546
  • Cas Number : 71025-82-6
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3597

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3597
Availability details for
Quantity
  • Availability
  • Add to Cart