COM448612620 Combi-Blocks, Inc.

N-(2-Aminophenyl)-N-ethylmethanesulfonamide

  • MDL number MFCD16679821
  • Cas Number : 869945-03-9
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3599

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3599
Availability details for
Quantity
  • Availability
  • Add to Cart