COM448612621 Combi-Blocks, Inc.

Benzyl N-[4-(2-aminoethyl)phenyl]carbamate

  • MDL number MFCD28096629
  • Cas Number : 1139884-44-8
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3600

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3600
Availability details for
Quantity
  • Availability
  • Add to Cart