COM448612623 Combi-Blocks, Inc.

3,5-Dimethyl-1-(4-nitrophenyl)pyrazole

  • MDL number MFCD00187810
  • Cas Number : 13788-94-8
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3607

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3607
Availability details for
Quantity
  • Availability
  • Add to Cart