COM448612638 Combi-Blocks, Inc.

1,3-Dibromo-4-fluoro-2-methyl-5-nitrobenzene

  • MDL number MFCD28127176
  • Cas Number : 1820613-94-2
  • Molecular Weight : 312.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3652
Availability details for
Quantity
  • Availability
  • Add to Cart