COM448612640 Combi-Blocks, Inc.

2-(Dimethoxymethyl)-4-methoxy-1-nitrobenzene

  • MDL number MFCD05148760
  • Cas Number : 548798-22-7
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3654

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3654
Availability details for
Quantity
  • Availability
  • Add to Cart