COM448612641 Combi-Blocks, Inc.

5-Bromo-3-chloro-2-fluoroaniline

  • MDL number MFCD28143920
  • Cas Number : 1517200-74-6
  • Molecular Weight : 224.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3655

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3655
Availability details for
Quantity
  • Availability
  • Add to Cart