COM448612643 Combi-Blocks, Inc.

2-Ethoxy-4-fluoroaniline

  • MDL number MFCD07801192
  • Cas Number : 178993-28-7
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3657
Availability details for
Quantity
  • Availability
  • Add to Cart