COM448612646 Combi-Blocks, Inc.

4-(Benzyloxy)-2-fluoroaniline

  • MDL number MFCD16154834
  • Cas Number : 190060-72-1
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3660

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3660
Availability details for
Quantity
  • Availability
  • Add to Cart