COM448612653 Combi-Blocks, Inc.

2,5-Diamino-3-bromobenzonitrile

  • MDL number MFCD22205744
  • Cas Number : 82997-65-7
  • Molecular Weight : 212.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3667
Availability details for
Quantity
  • Availability
  • Add to Cart