COM448612654 Combi-Blocks, Inc.

2,5-Diamino-3-chlorobenzonitrile

  • MDL number MFCD08704820
  • Cas Number : 82997-64-6
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3668
Availability details for
Quantity
  • Availability
  • Add to Cart