COM448612661 Combi-Blocks, Inc.

Ethyl 2-{[2-amino-4-(trifluoromethyl)phenyl]amino}acetate

  • MDL number MFCD12522986
  • Cas Number : 1178799-62-6
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3679
Availability details for
Quantity
  • Availability
  • Add to Cart