COM448612665 Combi-Blocks, Inc.

Benzyl N-(3-amino-5-bromophenyl)carbamate

  • MDL number MFCD22205746
  • Cas Number : 1375068-61-3
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3684
Availability details for
Quantity
  • Availability
  • Add to Cart