COM448612666 Combi-Blocks, Inc.

4-Methoxynaphthalen-1-amine

  • MDL number MFCD09743196
  • Cas Number : 16430-99-2
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3685

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3685
Availability details for
Quantity
  • Availability
  • Add to Cart