COM448612667 Combi-Blocks, Inc.

2,6-Dibromobenzene-1,4-diamine

  • MDL number MFCD02575406
  • Cas Number : 29213-03-4
  • Molecular Weight : 265.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3686
Availability details for
Quantity
  • Availability
  • Add to Cart