COM448612669 Combi-Blocks, Inc.

4,5-Dichloro-2-methylbenzene-1,3-diamine

  • MDL number MFCD20722348
  • Cas Number : 1375068-66-8
  • Molecular Weight : 191.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3688

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3688
Availability details for
Quantity
  • Availability
  • Add to Cart