COM448612674 Combi-Blocks, Inc.

4-Bromo-1-N-tert-butyl-5-fluorobenzene-1,2-diamine

  • MDL number MFCD21686296
  • Cas Number : 1375068-67-9
  • Molecular Weight : 261.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3693
Availability details for
Quantity
  • Availability
  • Add to Cart