COM448612676 Combi-Blocks, Inc.

3-Bromo-4-propoxyaniline

  • MDL number MFCD16085584
  • Cas Number : 156783-06-1
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3697

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3697
Availability details for
Quantity
  • Availability
  • Add to Cart