COM448612678 Combi-Blocks, Inc.

4-Bromo-5-propoxybenzene-1,2-diamine

  • MDL number MFCD22205748
  • Cas Number : 1375068-73-7
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3699

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3699
Availability details for
Quantity
  • Availability
  • Add to Cart