COM448612692 Combi-Blocks, Inc.

4-Bromo-5-ethoxy-1-N-propylbenzene-1,2-diamine

  • MDL number MFCD22192368
  • Cas Number : 1373233-24-9
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3714

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3714
Availability details for
Quantity
  • Availability
  • Add to Cart