COM448612697 Combi-Blocks, Inc.

4-Bromo-1-N-butyl-5-ethoxybenzene-1,2-diamine

  • MDL number MFCD22192372
  • Cas Number : 1373232-94-0
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3719

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3719
Availability details for
Quantity
  • Availability
  • Add to Cart