COM448612699 Combi-Blocks, Inc.

3-Amino-5-bromo-4-fluorobenzoic acid

  • MDL number MFCD22192374
  • Cas Number : 1290117-11-1
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3721
Availability details for
Quantity
  • Availability
  • Add to Cart