COM448612704 Combi-Blocks, Inc.

4-Bromo-6-(trifluoromethoxy)benzene-1,3-diamine

  • MDL number MFCD22192379
  • Cas Number : 1373233-29-4
  • Molecular Weight : 271.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3726
Availability details for
Quantity
  • Availability
  • Add to Cart