COM448612705 Combi-Blocks, Inc.

2-Butoxy-4-fluoroaniline

  • MDL number MFCD09739012
  • Cas Number : 954267-65-3
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3727
Availability details for
Quantity
  • Availability
  • Add to Cart