COM448612706 Combi-Blocks, Inc.

4-Fluoro-2-(pentyloxy)aniline

  • MDL number MFCD09811884
  • Cas Number : 1019114-95-4
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3728
Availability details for
Quantity
  • Availability
  • Add to Cart