COM448612710 Combi-Blocks, Inc.

5,6-Difluoro-1-N-methylbenzene-1,2-diamine

  • MDL number MFCD16787167
  • Cas Number : 1248508-73-7
  • Molecular Weight : 158.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3732
Availability details for
Quantity
  • Availability
  • Add to Cart