COM448612711 Combi-Blocks, Inc.

1-N-Benzyl-5-ethoxybenzene-1,2-diamine

  • MDL number MFCD22192380
  • Cas Number : 1373232-39-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3733
Availability details for
Quantity
  • Availability
  • Add to Cart