COM448612719 Combi-Blocks, Inc.

1-N-Butyl-5,6-difluorobenzene-1,2-diamine

  • MDL number MFCD16787174
  • Cas Number : 1249528-13-9
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3741
Availability details for
Quantity
  • Availability
  • Add to Cart