COM448612721 Combi-Blocks, Inc.

(4-Amino-2-fluorophenyl)acetic acid

  • MDL number MFCD09032899
  • Cas Number : 914224-31-0
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3743
Availability details for
Quantity
  • Availability
  • Add to Cart