COM448612733 Combi-Blocks, Inc.

(5-Amino-2-methylphenyl)boronic acid

  • MDL number MFCD22192389
  • Cas Number : 1003042-50-9
  • Molecular Weight : 151.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3756
Availability details for
Quantity
  • Availability
  • Add to Cart