COM448612736 Combi-Blocks, Inc.

[3-(4-Aminophenoxy)phenyl]boronic acid

  • MDL number MFCD22192393
  • Cas Number : 2096333-48-9
  • Molecular Weight : 229.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3763
Availability details for
Quantity
  • Availability
  • Add to Cart