COM448612737 Combi-Blocks, Inc.

{3-[2-Amino-4-(trifluoromethyl)phenoxy]phenyl}boronic acid

  • MDL number MFCD22192394
  • Cas Number : 2096336-78-4
  • Molecular Weight : 297.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3764
Availability details for
Quantity
  • Availability
  • Add to Cart