COM448612740 Combi-Blocks, Inc.

5-Amino-2-hydroxypyridine-3-carboxylic acid

  • MDL number MFCD17168425
  • Cas Number : 89640-79-9
  • Molecular Weight : 154.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3769
Availability details for
Quantity
  • Availability
  • Add to Cart