COM448612754 Combi-Blocks, Inc.

Tert-butyl 4-(4-amino-2-bromophenoxy)piperidine-1-carboxylate

  • MDL number MFCD22192399
  • Cas Number : 337520-17-9
  • Molecular Weight : 371.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3786

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3786
Availability details for
Quantity
  • Availability
  • Add to Cart