COM448612762 Combi-Blocks, Inc.

Tert-butyl 4-(5-aminoquinolin-8-yl)piperazine-1-carboxylate

  • MDL number MFCD22192401
  • Cas Number : 1373232-98-4
  • Molecular Weight : 328.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3794
Availability details for
Quantity
  • Availability
  • Add to Cart