COM448612765 Combi-Blocks, Inc.

6-N-Ethylquinoline-5,6-diamine

  • MDL number MFCD12067808
  • Cas Number : 1157504-04-5
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3798
Availability details for
Quantity
  • Availability
  • Add to Cart