COM448612766 Combi-Blocks, Inc.

6-(Pyrrolidin-1-yl)quinolin-5-amine

  • MDL number MFCD12067776
  • Cas Number : 21194-95-6
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3799

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3799
Availability details for
Quantity
  • Availability
  • Add to Cart